Learn More
PYREX™ Glass Extraction Thimbles Available on GSA/VA Contract for Federal Government customers only.
Resist attack by most chemicals
Manufacturer:  Corning 33950LEC
 View more versions of this product
Catalog No. 09-653D
Add to cart
Description
Description
  • Borosilicate glass
  • With fritted disc of coarse (nominal max. pore size, 40-60µm) or extra-coarse (nominal max. pore size, 170-220µm) porosity
  • Fit into standard Soxhlet extraction tubes
  • Convenient for weighing extracted materials
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Borosilicate Glass
4.5cm
For Pyrex extractors and extraction apparatus, soxhlet extractors
13cm
170 to 220μm
Extraction Thimble
1.77 in.
For weighing extracted materials
5.1 in.
Fritted discs
Extra coarse