PYREX™ Glass Manometers
Tube enclosed in round-bottom, graduated jacket and sealed to standard taper stopcock plug
Manufacturer: Corning, Inc.Supplier Diversity Partner 6950235
 View more versions of this product
Catalog No. 11-288
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Performing quality control functions including microbial detection, equipment calibration, method validation and bioburden determination
    Height (English) 9.25 in.
    Height (Metric) 23.5cm
    Includes Standard taper stopcock with 10mm O.D. Side tubulation arm
    Product Type Mercury Vacuum Manometer
  • Description & Specifications
Related Products
Product Content Feedback