Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Glass Pestle Tissue Grinders Available on GSA/VA Contract for Federal Government customers only.

Consist of glass tube and pestle with clearance of 0.15mm

Manufacturer:  Corning 772516

 View more versions of this product

Catalog No. 08-414B



Add to cart

Alert:

Operator should wear protective gloves with sufficient thickness to prevent injury in the event of failure of the glass components.


Description

Description

  • Parts ground to medium grind for abrasive action
  • Designed to be motor driven at low speed (provided that the attachment contains some form of friction release)
  • Rubber tubing can be placed on pestle for connection to chuck of drive motor

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

0.3 oz.
Borosilicate Glass
Designed for grinding small samples for pathological and biochemical work
150mm
Beaded rim
0.3 oz., 9mL
9mL
Tissue Grinder
5.9 in.
0.7 in.
18mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit