Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Globe-Shaped Funnels with standard taper Stopcock and Stopper Available on GSA/VA Contract for Federal Government customers only.

Excellent for rack work

Manufacturer:  Corning, Inc. 6340250

 View more versions of this product

Catalog No. 10-442C

  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

14 in.
8.4 oz.
3.3 in.
For rack work
Separatory Funnel
22
8.4 oz., 250mL
357mm
Glass
84.1mm
Stopper and stopcock
2
250mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit