Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Griffin Beakers Available on GSA/VA Contract for Federal Government customers only.

With spouts and strengthened rim with flare trimmed back that offer optimum balance between thermal shock resistance and mechanical strength

Manufacturer:  Corning, Inc. 100010

 View more versions of this product

Catalog No. 02-540C



  Check Availability
Add to cart

Description

Description

With spouts and strengthened rim with flare trimmed back that offer optimum balance between thermal shock resistance and mechanical strength

  • With pouring spout
  • Extra-large marking spot
  • Dual white scale

  • 10 to 50mL sizes suitable for microanalysis
  • 10mL size is not graduated
  • 250 through 4000mL capacity beakers have double graduated metric scale

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Compliance

Designed to meet ASTM™ E-960 requirements

Specifications

Specifications

0.33 oz.
0.98 in.
No
Yes
3.2cm
White marking
Dual white metric scale
Beaker
Borosilicate Glass
ASTM E960
2.5cm
For optimum balance between thermal shock resistance and mechanical strength
1.25 in.
Spout
12 Pack
Griffin
10mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit