Learn More
PYREX™ Hirsch Type, Fritted Disc
SCI_ED
Very useful in preparation work.
Manufacturer:  PYREX™ 3621075M/EMD
 View more versions of this product
Catalog No. S345925
Add to cart
Description
Description
Helpful when making separations where it is necessary to wash the pecipitate and redissolve with chemicals which would attack filter paper.
Specifications
Specifications
75mm
Pyrex Glass
ASTM E128
30mm
70mm
10 to 15μm
8mm
139 mm
Preparative work, or in making separations
Fritted discs
Medium
Hirsch Funnel