Learn More
Corning™ PYREX™ Hirsch Type, Fritted Disc
SCI_ED
Very useful in preparation work.
Supplier:  Corning™ 3621050C/EMD
 View more versions of this product
Catalog No. S345921
Add to Cart
Description
Description
Helpful when making separations where it is necessary to wash the pecipitate and redissolve with chemicals which would attack filter paper.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50mm
Pyrex Glass
ASTM E128
20mm
75mm
40 to 60μm
8mm
118 mm
Preparative work, or in making separations
Fritted discs
Coarse
Hirsch Funnel
Product Suggestions
Product Suggestions