Learn More
PYREX™ Large-Capacity Bottles Available on GSA/VA Contract for Federal Government customers only.
For distilled water and solutions
Manufacturer:  Corning, Inc. 15969L
 View more versions of this product
Catalog No. 02-887-1
  Check Availability
Add to cart
Description
Description
For distilled water and solutions
  • 2.5, 3.5-gallon sizes are conventional bottle shape
  • Five and 12 gallon sizes are similar to carboys
  • Necks are tooled to increase mechanical strength and for uniform stopper fit
  • Nonsterile
  • Withstand hot air (dry) or steam (wet) sterilization
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Bottles take No. 12 size stoppers (not included).
Specifications
Specifications
294.4 oz., 9000mL
294.4 oz.
18.7 in.
187mm
9000mL
7.3 in.
Borosilicate Glass
475mm