Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Large-Tip Opening Reusable Volumetric Pipets Available on GSA/VA Contract for Federal Government customers only.

Designed for schools and laboratories where Class A tolerances aren't required

Manufacturer:  Corning, Inc. 71021X

 View more versions of this product

Catalog No. 13-650-5


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.5mL
Pyrex™
ASTM E969
No
To deliver
12.51 in.
31.8cm
0.017 oz.
No
Black
±0.01mL
Volumetric Pipet
Wide
0.017 oz., 0.5mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit