Learn More
PYREX™ Microchemistry Kit Accessories - Bulb-type Immersion Mercury Thermometer
For use with Pyrex Microchemistry kit.
Manufacturer:  Corning 7712
Catalog No. 11-300-92
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
The products indicated contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Description
  • -10° to +150°C in 1°C subdivisions (14° to 272°F)
  • White back for easier reading
Specifications
Specifications
1°C
−10° to 150°C
150°C
−10°C
159mm
14° to 272°F
272°F
14°F
6.25 in.