Learn More
PYREX™ Microchemistry Kit Accessories - Syringe
Replacement part for PYREX Microchemistry Kits
Manufacturer:  Corning 93501
Catalog No. 11-300-96
Add to cart
Description
Description
  • 1cc capacity, in 0.01cc graduations
  • Glass barrel
  • TFE tip plunger
  • Without needle
Order Info
For 11-300-71 and 11-300-74 Microchemistry Kits
Specifications
Specifications
0.03 oz.
Syringe
Glass tip plunger
0.03 oz., 1mL
1mL
For PYREX™ microchemistry kit
Glass