Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Pharmaceutical Beakers Available on GSA/VA Contract for Federal Government customers only.

With double spout and double scale

Manufacturer:  Corning, Inc. 6480125

 View more versions of this product

Catalog No. 02-558A



  Check Availability
Add to cart

Alert:

Do not subject directly to heat source.


Description

Description

With double spout and double scale

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

4.22 oz.
5.6cm
Ideal for measuring hot solutions
5 to 125mL
10.2cm
5mL
1 Pack
Beaker
Borosilicate Glass
2.2 in.
No
Yes
4.01 in.
Double spout
Marking spot
Dual white scale
125mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit