PYREX™ Polypropylene Flask Closures Available on GSA/VA Contract for Federal Government customers only.

Designed for culture tubes and flasks with long rimless necks

Manufacturer: Corning, Inc. 2679638

 View more versions of this product

Catalog No. 14-957-85H

  Check Availability
Add to Cart

    Specifications

    Product Type Cap, Flask
    Diameter (Metric) Outer Neck 3.8cm
    Disposable No
    For Use With (Equipment) Culture tubes and flasks with long rimless necks
    Sterile No
    Closure Color Orange
    Autoclavable Yes
    Closure Material Polypropylene
    Diameter (English) Outer Neck 1.49 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit