Learn More
PYREX™ Reusable Class A Volumetric Serialized/Certified Pipets Available on GSA/VA Contract for Federal Government customers only.
Borosilicate-glass with fire-polished top and tip
Manufacturer:  Corning, Inc. 710110
 View more versions of this product
Catalog No. 13-651-11E
  Check Availability
Add to cart
Alert:
Do not pipet by mouth. Use mechanical pipetting device
Specifications Description & Specifications
Specifications
Yes
Pyrex™
ASTM E542, ASTM E969, certificate of identification and capacity, NIST traceable
Red
±0.02mL
A
Mechanical pipetting device
Round
10mL
No
Yes
No
To deliver
Serialized Volumetric Pipet
16.88 in.
42.9cm