Learn More
PYREX™ Reusable Class A Volumetric Serialized/Certified Pipets Available on GSA/VA Contract for Federal Government customers only.
Borosilicate-glass with fire-polished top and tip
Manufacturer:  Corning, Inc. 71012
 View more versions of this product
Catalog No. 13-651-11A
  Check Availability
Add to cart
Alert:
Do not pipet by mouth. Use mechanical pipetting device
Specifications Description & Specifications
Specifications
2mL
No
Orange
±0.01mL
A
Mechanical pipetting device
Round
Pyrex™
ASTM E542, ASTM E969, certificate of identification and capacity, NIST traceable
No
To deliver
Serialized Volumetric Pipet
13.11 in.
33.3cm