Learn More
PYREX™ Reusable Class A Volumetric Serialized/Certified Pipets Available on GSA/VA Contract for Federal Government customers only.
Borosilicate-glass with fire-polished top and tip
Manufacturer:  Corning, Inc. 71014
 View more versions of this product
Catalog No. 13-651-11C
  Check Availability
Add to cart
Alert:
Do not pipet by mouth. Use mechanical pipetting device
Specifications Description & Specifications
Specifications
4mL
No
Red
±0.01mL
A
Mechanical pipetting device
Round
Pyrex™
ASTM E542, ASTM E969, certificate of identification and capacity, NIST traceable
No
To deliver
Serialized Volumetric Pipet
14.56 in.
37cm