Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Reusable Glass Tubes with Rubber-Lined Phenolic Caps

Deep-form phenolic caps facilitate handling and sealing after autoclaving

Supplier:  Corning™ 982525X

 View more versions of this product

Catalog No. 14-932F



Add to Cart

Description

Description

Deep-form phenolic caps facilitate handling and sealing after autoclaving

Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. Caps are provided with a glued-in, white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations. Pencil marks are easily erased.

For phenolic caps, see Cat. No. 9999.

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
Threaded
200 mm
Marking Spot
24-410
Autoclavable
25 mm
Culture Tube
Phenolic
70 mL
Borosilicate Glass
Rubber
Non-sterile
No
Screw Cap
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit