PYREX™ Reusable Glass Tubes with Rubber-Lined Caps Available on GSA/VA Contract for Federal Government customers only.

Deep-form phenolic caps facilitate handling and sealing after autoclaving

Manufacturer: Corning, Inc. 982516X

 View more versions of this product

Catalog No. 14-932A

  Check Availability
Add to Cart

    Specifications

    Closure Color Black
    Closure Material Phenolic
    Design Type Threaded
    Graduated No
    Label Type Marking Spot
    Liner Material Rubber
    Thread Size 15-415
    Capacity (Metric) 15mL
    Closure Type Screw
    Diameter (Metric) Outer 16mm
    Length (Metric) 125mm
    Material Borosilicate Glass
    Shape Round Bottom
    Sterile No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit