Learn More
PYREX™ Reusable Petri Dishes: Bottoms Only Available on GSA/VA Contract for Federal Government customers only.
For use with Corning PYREX Brand Reusable Petri Dishes
Manufacturer:  Corning 3160101BO
 View more versions of this product
Catalog No. 08-748C
Add to cart
Description
Description
  • Withstand repeated sterilization
  • Not affected chemically or thermally
  • Bottoms marked in white with triangular enamel reference point for serial dilutions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100 mm
15mm
08-747C
Pyrex
Replacement Bottom
Beaded