PYREX™ Reusable Test Tubes with Beaded Rim Available on GSA/VA Contract for Federal Government customers only.

Sizes from 10 to 20mm O.D. have marking spot

Manufacturer: Corning, Inc. 980015

 View more versions of this product

Catalog No. 14-955D

  Check Availability
Add to Cart

    Specifications

    Design Type Plain
    Graduated No
    Label Type Marking Spot
    Capacity (Metric) 14mL
    Diameter (Metric) Outer 15mm
    Length (Metric) 125mm
    Material Borosilicate Glass
    Shape Cylindrical, Round Bottom
    Sterile No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit