Learn More
PYREX™ Round-Bottom Glass Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Without graduations
Manufacturer:  Corning, Inc. 824050
 View more versions of this product
Catalog No. 05-550
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
29mm
50mL
Borosilicate Glass
Useful for the handling of sputum specimens, digestion, shaking, neutralizing and centrifuging
116mm
No
No
Round with Pourout Top