Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lenz Laborglasinstrumente™ PYREX™ Serialized and Certified Flasks, Low Actinic Available on GSA/VA Contract for Federal Government customers only.

Featuring red low actinic stained glass to provide protection for materials sensitive to light. Lenz Laborglasinstrumente™ PYREX™ Serialized and Certified Flasks, Low Actinic are tested and calibrated in an ISO/IEC 17025 certified laboratory.

Manufacturer:  Lenz Laborglasinstrumente™ 556801L

 View more versions of this product

Catalog No. 11-850-25


  Check Availability
Add to cart

Specifications

Description

  • Sharp graduation line
  • With inscription label
  • Calibrated "to contain" within Class A tolerances
  • Corrosion-free and resistant to thermal shock
Description & Specifications

Specifications

Flat
Glass
4.9 in.
Yes
34.4cm
±0.3mL
Glass,Pyrex™
Calibrated To Contain
22
125mm
13.54 in.
Long
1000mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit