Learn More
PYREX™ Shorty Disposable Glass Serological Pipets Available on GSA/VA Contract for Federal Government customers only.
Ideal for use in confined areas such as hoods
Manufacturer:  Corning, Inc. 7078B1
 View more versions of this product
Catalog No. 13-664-18A
  Check Availability
Add to cart
Specifications
Description
  • Sterile, plugged pipets have tooled tops
  • Negative graduations make it applicable for both serological and measuring procedures
  • I.S.O. color coding is printed directly on Steri-View™ (one side paper, one side plastic) wrapper
Description & Specifications
Specifications
8.4 in.
0.5mL
1mL
Pyrex™
Serological Pipet
Round
0.03 oz., 1mL
Yes
215mm
0.03 oz.
Yes
Yellow
Yes
0.01mL
Yes
Multi-pack