PYREX™ Squibb Glass Separatory Funnels with Screw Cap and PTFE Valve Available on GSA/VA Contract for Federal Government customers only.

Constructed of durable PYREX borosilicate glass

Manufacturer: Corning, Inc. 630560

 View more versions of this product

Catalog No. 10-462-581

  Check Availability
Add to Cart

    Specifications

    Product Type Squibb Separatory Funnel
    Capacity (English) 2.02 oz.
    Quantity 1 Case
    Capacity (Metric) 60mL
    Capacity 2.02 oz., 60mL
    Material Pyrex™

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit