Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Squibb Pear-Shaped Separatory Funnels with Rotaflo PTFE-Plug Stopcock

Pear-shaped with standard taper ground neck, offset PTFE-plug stopcock and all-glass PYREX stopper

Manufacturer:  Corning, Inc. 6302500

 View more versions of this product

Catalog No. 10-437-6C


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.39 in.
15.1 in.
2.5 in.
Pyrex™
3.8 in.
For replacement glass stoppers
Each
27
16.9 oz., 500mL
1cm
38.5cm
65mm
16.9 oz.
9.8cm
Squibb Separatory Funnel
Polyethylene
500mL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit