Learn More
Corning™ PYREX™ Pear-Shaped Squibb Separatory Funnel with PTFE Product Standard Stopcock and Polyethylene Standard Taper Stopper
GREENER_CHOICE
With closed-bottom stoppers of linear polyethylene.
$291.00 - $591.00
Specifications
Material Pyrex Glass
For Use With (Application) For replacement polyethylene standard taper stoppers and PTFE stopcock assemblies
Length (Metric) Stem 65 mm
Stopper Material Polyethylene
Product Type Squibb separatory funnel
View More Specs
Products 6
Catalog Number Mfr. No. Height (Metric) Stopcock Stopper Number Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Stopcock Stopper Number Price Quantity & Availability  
10-437-5B
GREENER_CHOICE
View Documents
Corning™
6404125
28.9 cm 2; PTFE 22
Each for $297.00
Case of 4 EA for $829.00
Add to Cart
10-437-5A
View Documents
Corning™
640460
24.4 cm 2; PTFE 16
Each for $291.00
Case of 4 EA for $814.00
Add to Cart
10-437-5D
View Documents
Corning™
6404500
35.4 cm 4; PTFE 27
Each for $408.00
Case of 4 EA for $1,139.00
Add to Cart
10-437-5C
GREENER_CHOICE
View Documents
Corning™
6404250
31.1 cm 4; PTFE 22
Each for $356.00
Case of 4 EA for $994.00
Add to Cart
10-437-5F
GREENER_CHOICE
View Documents
Corning™
64042L
48.6 cm 6; PTFE 38
Case for $591.00
Add to Cart
10-437-5E
GREENER_CHOICE
View Documents
Corning™
64041L
39.2 cm 4; PTFE 27
Case for $408.00
Add to Cart
Description
Description
The stopper is made with a closed bottom and is of linear high density polyethylene to conform to stopper dimensions.
For stopper only, see Cat. No. 7624.
Order Info
Stem lengths, 65mm
Specifications
Specifications
Pyrex Glass
65 mm
Squibb separatory funnel
For replacement polyethylene standard taper stoppers and PTFE stopcock assemblies
Polyethylene