Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Squibb Separatory Funnel, PTFE Standard Stopcock, Glass Standard Taper Stopper Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  Corning, Inc. 64021L

 View more versions of this product

Catalog No. 10-437-2E


  Check Availability
Add to cart

Specifications

Description

Separatory funnels are used in liquid-liquid extractions to separate the components of a mixture into two immiscible solvent phases of different densities.
Description & Specifications

Specifications

2.5 in.
Pyrex™
33.8 oz.
11.9cm
Squibb Separatory Funnel
Glass
1L
65mm
4
4.6 in.
For replacement glass standard taper stoppers
1 Case
27
33.8 oz., 1L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit