Learn More
PYREX™ Squibb Separatory Funnel Without Valve And Stopper
Economy separatory funnel replacements
Manufacturer:  PYREX™ 64061LFO
Catalog No. 10-444-2
Description
Description
These PYREX economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper. For replacement polyethylene Standard Taper stoppers, see No. 7624; for replacement PTFE locking stopcock assemblies, see No. 2116-LSO; for replacement tips, see No. 6406-GTO.
Specifications
Specifications
1 L
Pyrex Glass
1/Cs.
27
For replacement polyethylene standard taper stoppers, for replacement tips
Replacement Squibb Separatory Funnel
PTFE