Learn More
PYREX™ Vacuum-Type Distillation Traps
Separable, with standard taper joints
$551.50 - $1662.00
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Outer | Standard Taper Joint No. | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Outer | Standard Taper Joint No. | Price | Quantity & Availability | |||
11-184-1B
|
PYREX™
772932 |
1.26 in. | 32 mm | 34/45 Inner Bottom, 29/42 Outer Top |
Case of 4 for $1,662.00
|
|
11-184-1E
|
PYREX™
772948 |
1.88 in. | 48 mm | 50/50 |
Case for $551.50
|
|
11-184-1D
|
PYREX™
772941 |
2.8 in. | 42 mm | 45/50 |
Case of 2 for $656.50
|
|
Description
With a full length outer joint on the outer tube and an inner joint on the inner tube. The top and side tubulations are the same O.D. as the center tube and have a length of 75 mm.
Specifications
Vacuum Trap | |
Pyrex™ |