Learn More
PYREX VISTA Micro Filter Flask with Sidearm Tubulation
Small flasks recommended for microchemical use
$199.40 - $208.50
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
07-250-093
|
PYREX
7036025 |
25 mL |
Pack of 6 for $199.40
Case of 2 PK for $296.50 |
|
07-250-094
|
PYREX
7036050 |
50 mL |
Pack of 6 for $200.50
Case of 2 PK for $299.50 |
|
07-250-095
![]() |
PYREX
70360125 |
125 mL |
Pack of 6 for $208.50
Case of 2 PK for $310.00 |
|
Description
Small flasks recommended for microchemical use
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots. The filtering flasks are designed to ASTM E-1406 Type 2.
Compliance
Designed to ASTM E-1406 Type 2
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Borosilicate Glass |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.