Learn More
PYREX™ VISTA™ Reusable Glass Tubes with Phenolic Screw Caps Available on GSA/VA Contract for Federal Government customers only.
Supplied with deep-form phenolic caps to facilitate handling and sealing after autoclaving
Manufacturer:  Corning 7082525
 View more versions of this product
Catalog No. 07-250-139
Add to cart
Description
Description
  • Blue enamel marking spot provides excellent surface for pencil notations
  • Pencil marks are easily erased
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Replacement caps available.
Specifications
Specifications
Phenolic
55mL
25mm
Screw
150mm
No
Blue Enamel Marking Spot
Round
24-410
Non-sterile
Threaded
Yes
Black
Borosilicate Glass
No
Rubber
Untreated