Learn More
PYREXPLUS™ Volumetric Flasks Available on GSA/VA Contract for Federal Government customers only.
PVC coating on the outside reduces chance of breakage
Manufacturer:  Corning, Inc. 65640250
 View more versions of this product
Catalog No. 10-240E
  Check Availability
Add to cart
Alert:
Do not place over direct heat or flame. Do not heat above 121°C (250°F) with moist heat or 110°C (230°F) dry heat.
Specifications Description & Specifications
Specifications
Flat
16
78mm
25.2cm
±0.12mL
Glass,Pyrexplus™
Glass
3.07 in.
9.92 in.
Long
250mL