Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridazine, 98+%, Thermo Scientific Chemicals

Catalog Number p-189610
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
289-80-5
C4H4N2
80.09
MFCD00006463
PBMFSQRYOILNGV-UHFFFAOYSA-N
orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
9259
CHEBI:30954
pyridazine
C1=CC=NN=C1
289-80-5
C4H4N2
80.09
MFCD00006463
PBMFSQRYOILNGV-UHFFFAOYSA-N
orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
9259
CHEBI:30954
pyridazine
C1=CC=NN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -8°C
Color Yellow to Brown
Density 1.1030g/mL
Boiling Point 208°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5225 to 1.5245
Quantity 5 g
Beilstein 23,89
Specific Gravity 1.103
Merck Index 15,808
Solubility Information Solubility in water: miscible.,miscible with benzene and dmf,freely soluble in methanol,ethanol and ether,practically insoluble in petroleum ether
Formula Weight 80.09
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Pyridazine, 98+%
Show More Show Less
EINECSNumber 206-025-5
RTECSNumber UR5267000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit