Alfa Aesar™ Pyridine-2-carboxaldehyde 2-quinolylhydrazone, 98+%

Manufacturer: Alfa Aesar™ B2312806

Quantity 5g

 View more versions of this product

Catalog No. AAB2312806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7385-99-1
    Molecular Formula C15H12N4
    Formula Weight 248.29
    MDL Number MFCD00014660
    Beilstein 480209
    Synonym PAQU
    Chemical Name or Material Pyridine-2-carboxaldehyde 2-quinolylhydrazone

    Specifications

    Formula Weight 248.29
    TSCA TSCA
    Molecular Formula C15H12N4
    MDL Number MFCD00014660
    CAS 7385-99-1
    Beilstein 480209
    Synonym PAQU
    Melting Point 203° to 206°C
    Quantity 5g
    Chemical Name or Material Pyridine-2-carboxaldehyde 2-quinolylhydrazone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit