Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone, 98%

Click to view available options
Quantity:
5g
25g
70421-66-8
C12H10N4O2
242.238
MFCD00012100
JXFHYUCJPGWCFI-NTEUORMPSA-N
pyridine-2-carboxaldehyde 4-nitrophenylhydrazone,2-pyridinecarboxaldehyde 4-nitrophenylhydrazone,pyridine-2-carbaldehyde 4'-nitrophenylhydrazone,2-4-nitrobenzenecarbohydrazonoyl pyridine,picolinaldehyde 4-nitrophenylhydrazone,pyridine-2-carbaldehyde 4-nitrophenylhydrazone,4-nitro-n-e-2-pyridinylmethylideneamino aniline,4-nitro-n-e-pyridin-2-ylmethylideneamino aniline,1e-2-2-pyridyl-1-azavinyl 4-nitrophenyl amine
9562617
4-nitro-N-[(E)-pyridin-2-ylmethylideneamino]aniline
C1=CC=NC(=C1)C=NNC2=CC=C(C=C2)[N+](=O)[O-]
70421-66-8
C12H10N4O2
242.238
MFCD00012100
JXFHYUCJPGWCFI-NTEUORMPSA-N
pyridine-2-carboxaldehyde 4-nitrophenylhydrazone,2-pyridinecarboxaldehyde 4-nitrophenylhydrazone,pyridine-2-carbaldehyde 4'-nitrophenylhydrazone,2-4-nitrobenzenecarbohydrazonoyl pyridine,picolinaldehyde 4-nitrophenylhydrazone,pyridine-2-carbaldehyde 4-nitrophenylhydrazone,4-nitro-n-e-2-pyridinylmethylideneamino aniline,4-nitro-n-e-pyridin-2-ylmethylideneamino aniline,1e-2-2-pyridyl-1-azavinyl 4-nitrophenyl amine
9562617
4-nitro-N-[(E)-pyridin-2-ylmethylideneamino]aniline
C1=CC=NC(=C1)C=NNC2=CC=C(C=C2)[N+](=O)[O-]
Quantity 25g
Melting Point 256°C to 259°C
Formula Weight 242.24
Percent Purity 98%
Chemical Name or Material Pyridine-2-carboxaldehyde 4-nitrophenylhydrazone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning