Alfa Aesar™ Pyridine-2-carboxaldoxime, 99%

Manufacturer: Alfa Aesar™ A1671436

Quantity 500g

 View more versions of this product

Catalog No. AAA1671436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 873-69-8
    Molecular Formula C6H6N2O
    Formula Weight 122.13
    MDL Number MFCD00006354
    Beilstein 109061
    Synonym Pyridine-2-aldoxime
    Chemical Name or Material Pyridine-2-carboxaldoxime

    Specifications

    Formula Weight 122.13
    Molecular Formula C6H6N2O
    MDL Number MFCD00006354
    CAS 873-69-8
    Synonym Pyridine-2-aldoxime
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    RTECS Number TJ4180000
    Melting Point 110° to 112°C
    Quantity 500g
    Beilstein 109061
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Pyridine-2-carboxaldoxime
    Assay 99%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit