Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyridine-2-carboxaldoxime methochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
51-15-0
C7H9ClN2O
172.612
MFCD00011981
HIGSLXSBYYMVKI-NAFXZHHSSA-N
pralidoxime chloride,1-methylpyridinium-2-aldoxime chloride,2-pam-cl,2-hydroxyimino methyl-1-methylpyridin-1-ium chloride,2-hydroxyimino methyl-1-methylpyridinium chloride,2-formyl 1-methyl pyridinium oxime chloride
57428996
[(Z)-(1-methylpyridin-2-ylidene)methyl]-oxoazanium;chloride
CN1C=CC=CC1=C[NH+]=O.[Cl-]
51-15-0
C7H9ClN2O
172.612
MFCD00011981
HIGSLXSBYYMVKI-NAFXZHHSSA-N
pralidoxime chloride,1-methylpyridinium-2-aldoxime chloride,2-pam-cl,2-hydroxyimino methyl-1-methylpyridin-1-ium chloride,2-hydroxyimino methyl-1-methylpyridinium chloride,2-formyl 1-methyl pyridinium oxime chloride
57428996
[(Z)-(1-methylpyridin-2-ylidene)methyl]-oxoazanium;chloride
CN1C=CC=CC1=C[NH+]=O.[Cl-]
Melting Point ~223°C (decomposition)
Quantity 100g
Beilstein 4163981
Merck Index 14,7703
Formula Weight 172.62
Percent Purity 97%
Chemical Name or Material Pyridine-2-carboxaldoxime methochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UU4200000