Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyridine-2-carboxamide, 98%

Click to view available options
Quantity:
5g
25g
1452-77-3
C6H6N2O
122.127
MFCD00023483
IBBMAWULFFBRKK-UHFFFAOYSA-N
picolinamide,2-pyridinecarboxamide,picolinoylamide,2-carbamoylpyridine,picolinic acid amide,2-picolinamide,2-aminocarbonyl-pyridine,alpha-picolinamide,pyridinecarboxamide,unii-i3550ccl59
15070
CHEBI:8200
pyridine-2-carboxamide
C1=CC=NC(=C1)C(=O)N
1452-77-3
C6H6N2O
122.127
MFCD00023483
IBBMAWULFFBRKK-UHFFFAOYSA-N
picolinamide,2-pyridinecarboxamide,picolinoylamide,2-carbamoylpyridine,picolinic acid amide,2-picolinamide,2-aminocarbonyl-pyridine,alpha-picolinamide,pyridinecarboxamide,unii-i3550ccl59
15070
CHEBI:8200
pyridine-2-carboxamide
C1=CC=NC(=C1)C(=O)N
Boiling Point 143°C (20mmHg)
Quantity 5g
Melting Point 104°C to 108°C
Beilstein 109617
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material Pyridine-2-carboxamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 215-921-5

TSCA : Yes

Recommended Storage : Ambient temperatures