Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-3-boronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
1692-25-7
C5H6BNO2
122.92
MFCD00674177
ABMYEXAYWZJVOV-UHFFFAOYSA-N
pyridine-3-boronic acid,3-pyridylboronic acid,3-pyridineboronic acid,pyridin-3-yl boronic acid,3-pyridinylboronic acid,pyridin-3-yl-3-boronic acid,3-pyridineboric acid,3-pyridinebornic acid,pyridyl-3-boronic acid,pyridine 3-boronic acid
2734378
pyridin-3-ylboronic acid
B(C1=CN=CC=C1)(O)O
1692-25-7
C5H6BNO2
122.92
MFCD00674177
ABMYEXAYWZJVOV-UHFFFAOYSA-N
pyridine-3-boronic acid,3-pyridylboronic acid,3-pyridineboronic acid,pyridin-3-yl boronic acid,3-pyridinylboronic acid,pyridin-3-yl-3-boronic acid,3-pyridineboric acid,3-pyridinebornic acid,pyridyl-3-boronic acid,pyridine 3-boronic acid
2734378
pyridin-3-ylboronic acid
B(C1=CN=CC=C1)(O)O
Packaging Glass bottle
Quantity 1g
Color Undesignated
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Formula Weight 122.92
Physical Form Powder, Crystals and/or Chunks
Percent Purity 95%
Chemical Name or Material Pyridine-3-boronic acid

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning