Pyridine, anhydrous, >99.5%, Alfa Aesar™

Manufacturer: Alfa Aesar 43799K2

Quantity 1L

 View more versions of this product

Catalog No. AA43799K2

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 110-86-1
    Physical Form Liquid
    Assay Percent Range >99.5%
    Molecular Formula C5H5N
    Formula Weight 79.1
    MDL Number MFCD00011732
    Chemical Name or Material Pyridine

    Specifications

    Formula Weight 79.1
    Physical Form Liquid
    Color Clear colorless
    Quantity 1L
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 203-809-9
    Ignition Point 482°C (900°F)
    MDL Number MFCD00011732
    CAS 110-86-1
    Odor Unpleasant
    Vapor Pressure 15mm Hg at 20°C
    Assay Percent Range >99.5%
    Molecular Formula C5H5N
    Health Hazard 1 H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
    Chemical Name or Material Pyridine

    H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit