Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
500mL
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

110-86-1
C5H5N
79.102
JUJWROOIHBZHMG-UHFFFAOYSA-N
1049
CHEBI:16227

High quality chemicals for laboratory and specialized industrial use

CAS 110-86-1
Molecular Formula C5H5N
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.98kg
Flash Point 20.00°C
Quantity 500mL
Chemical Name or Material Pyridine
Grade Baker Analyzed™ ACS Reagent
Show More Show Less