Alfa Aesar™ Pyridine-d5, 99.5%(Isotopic)

Manufacturer: Alfa Aesar™ 42341SA

Quantity 2 each

 View more versions of this product

Catalog No. AA42341SA

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7291-22-7
    Formula Weight 84.14
    MDL Number MFCD00044639
    UN Number UN1282
    Beilstein 114377
    Chemical Name or Material Pyridine-d5, Isotopic

    Specifications

    Formula Weight 84.14
    Packaging Prescored Ampules
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.05g/mL
    Boiling Point 115°C
    Flash Point 20°C (68°F)
    MDL Number MFCD00044639
    UN Number UN1282
    Health Hazard 2 GHS H Statement
    H225-H302-H312-H332
    Highly flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Physical Form Liquid
    Quantity 2 each
    Health Hazard 1 Danger
    Odor Unpleasant
    Melting Point −42°C
    Refractive Index 1.506
    CAS 7291-22-7
    Beilstein 114377
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
    Chemical Name or Material Pyridine-d5, Isotopic
    Assay 99.5%

    GHS H Statement
    H225-H302-H312-H332
    Highly flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit