Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-d5, for NMR, 100.0% atom D, ACROS Organics™

Click to view available options
Quantity:
1mL
10mL
25mL
Packaging:
Glass bottle
7291-22-7
C5H5N
84.13
MFCD00044639
JUJWROOIHBZHMG-RALIUCGRSA-N
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ~2~h_5_ pyridine, de85c
558519
2,3,4,5,6-pentadeuteriopyridine
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
7291-22-7
C5H5N
84.13
MFCD00044639
JUJWROOIHBZHMG-RALIUCGRSA-N
pyridine-d5, 2h5 pyridine, c5d5n, 2 h? pyridine, pyridine d5, pyridine,crude,light, pyridine, perdeutero, ~2~h_5_ pyridine, de85c
558519
2,3,4,5,6-pentadeuteriopyridine
[2H]C1=NC([2H])=C([2H])C([2H])=C1[2H]
Color Colorless
Deuterium (D) 99.95% min.
Packaging Glass bottle
Water 0.02% max. (Coulometric)
Density 1.0500g/mL
Quantity 1mL
Boiling Point 114.4°C
Melting Point -41.0°C
Flash Point 20°C
Beilstein 20, III, 2305
Refractive Index 1.506
Formula Weight 84.14
Physical Form Liquid
Grade NMR
Chemical Name or Material Pyridine-d5, 100 atom%D
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger