Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyridine N-oxide, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Pyridine N-oxide is used in the treatment of Kayser's disease. It is widely used in chemical industry. It can act as catalytic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
Soluble in water.
Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
Specifications
Melting Point | 61°C to 66°C |
Boiling Point | 270°C |
Flash Point | 143°C (289°F) |
Quantity | 25 g |
Beilstein | 105257 |
Sensitivity | Very Hygroscopic |
Merck Index | 14,7972 |
Solubility Information | Soluble in water. |
Formula Weight | 95.1 |
Percent Purity | 95% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P261-P280-P301+P310-P305+P351+P338-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 211-774-6 |
RTECSNumber | UT6410000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction