Pyridine, Ultrapure, Spectrophotometric Grade, >99.5%, Alfa Aesar™

Manufacturer: Alfa Aesar 32436K7

Quantity 4L

 View more versions of this product

Catalog No. AA32436K7

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 110-86-1
    Physical Form Liquid
    Assay Percent Range >99.5%
    Molecular Formula C5H5N
    Formula Weight 79.1
    MDL Number MFCD00011732
    Chemical Name or Material Pyridine
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 79.1
    Quantity 4L
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: II
    EINECS Number 203-809-9
    Physical Form Liquid
    Vapor Pressure 20 hPa (15mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >99.5%
    Health Hazard 1 H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
    Odor Unpleasant
    Molecular Formula C5H5N
    MDL Number MFCD00011732
    CAS 110-86-1
    Chemical Name or Material Pyridine
    Grade Spectrophotometric grade

    H225-H302-H312-H332: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit