Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrido[2,3-b]pyrazine 99%, ACROS Organics™

Click to view available options
Quantity:
1g
322-46-3
C7H5N3
131.138
MFCD00006717
YEYHFKBVNARCNE-UHFFFAOYSA-N
pyrido 2,3-b pyrazine,1,4,5-triazanaphthalene,5-azaquinoxaline,pyrido 2,3 pyrazine,pyrido 3,2-b pyrazine,acmc-20ahgw,1,5-triazanaphthalene,pyridino 2,3-b pyrazine
67580
pyrido[2,3-b]pyrazine
C1=CC2=NC=CN=C2N=C1
322-46-3
C7H5N3
131.138
MFCD00006717
YEYHFKBVNARCNE-UHFFFAOYSA-N
pyrido 2,3-b pyrazine,1,4,5-triazanaphthalene,5-azaquinoxaline,pyrido 2,3 pyrazine,pyrido 3,2-b pyrazine,acmc-20ahgw,1,5-triazanaphthalene,pyridino 2,3-b pyrazine
67580
pyrido[2,3-b]pyrazine
C1=CC2=NC=CN=C2N=C1
Quantity 1g
Melting Point 139°C to 143°C
Color Brown to Green
Infrared Spectrum Authentic
Assay Percent Range 99%
Formula Weight 131.14
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Pyrido[2, 3-b]pyrazine, 99%

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.