Alfa Aesar™ Pyridoxal-5-phosphate hydrate, 98%

Manufacturer: Alfa Aesar™ A1232314

Quantity 25g

 View more versions of this product

Catalog No. AAA1232314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41468-25-1
    Molecular Formula C8H10NO6P·H2O
    Formula Weight 265.17 (247.15 Anhydrous)
    MDL Number MFCD00149414
    Merck Index 14,7979
    Chemical Name or Material Pyridoxal-5-phosphate hydrate

    Specifications

    Formula Weight 265.17 (247.15 Anhydrous)
    MDL Number MFCD00149414
    CAS 41468-25-1
    RTECS Number UV1207000
    TSCA TSCA
    Molecular Formula C8H10NO6P·H2O
    Merck Index 14,7979
    Recommended Storage −30°C to −10°C
    Melting Point 142°C
    Quantity 25g
    Chemical Name or Material Pyridoxal-5-phosphate hydrate
    Assay 98%

    Description

    Applications
    Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.

    Solubility
    Soluble in alkaline solutions. Sparingly soluble in water.

    Notes
    Light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.

    −30°C to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit