Learn More
Medchemexpress LLC Pyridoxal 5'-phosphate monohydrate | 41468-25-1 | 98.0% | 265.16 | 5G
SDP
Supplier:  Medchemexpress LLC HYW011727A5G
Encompass_Preferred
Pyridoxal 5'-phosphate monohydrate is the active form of vitamin B6, essential for multiple enzymes including aromatic l-amino acid decarboxylase, which produces neurotransmitters like dopamine and serotonin. It's a key coenzyme in vitamin B6 phosphorylation, interconvertible with pyridoxine 5′-phosphate (PNP) and pyridoxamine 5′-phosphate (PMP).
  • Active form of vitamin B6; essential cofactor for enzymes.
  • Crucial for dopamine and serotonin production.
  • Interconvertible coenzyme variant.
  • Purity: 98.0%.
  • Activates ERK/c-Jun signaling pathway.
  • Inhibits cancer cell proliferation (SK-OV-3, OVCAR-3).
  • Alleviates memory impairment (mouse models).
  • Prevents diabetic nephropathy progression (rat models).
  • Available as solid or in solutions.
  • For research use only.
Catalog No. 50-004-54062