Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
500g
Packaging:
Glass bottle
65-22-5
C8H9NO3·HCl
203.62
MFCD00012809
FCHXJFJNDJXENQ-UHFFFAOYSA-N
pyridoxal hydrochloride,pyridoxal hcl,vitamin b6 hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride,unii-1416kf0qbc,2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride,pl hcl,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride,3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
6171
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
CC1=NC=C(C(=C1O)C=O)CO.Cl
65-22-5
C8H9NO3·HCl
203.62
MFCD00012809
FCHXJFJNDJXENQ-UHFFFAOYSA-N
pyridoxal hydrochloride,pyridoxal hcl,vitamin b6 hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride,unii-1416kf0qbc,2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride,pl hcl,3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride,3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride,3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
6171
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
CC1=NC=C(C(=C1O)C=O)CO.Cl
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
Color White
Melting Point 171.0°C to 175.0°C
Assay Percent Range 99%
Merck Index 15,8091
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: soluble in 95% ethanol: 1.7g/100 mL
Formula Weight 203.62
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Pyridoxal hydrochloride
Show More Show Less

EINECSNumber : 200-602-5