Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxal Hydrochloride, Fisher BioReagents

Click to view available options
Quantity:
1g
5g
100g
Packaging:
Amber Glass
65-22-5
C8H10ClNO3
203.622
FCHXJFJNDJXENQ-UHFFFAOYSA-N
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
6171
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
CC1=NC=C(C(=C1O)C=O)CO.Cl
65-22-5
C8H10ClNO3
203.622
FCHXJFJNDJXENQ-UHFFFAOYSA-N
pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
6171
3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
CC1=NC=C(C(=C1O)C=O)CO.Cl
Pyridoxal Hydrochloride is useful for labeling amino acids and for their detection in the picomole range.
Packaging Amber Glass
Color White
pH 2.1
Quantity 1g
Identification Pass Test
Melting Point 171°C
Merck Index 15, 8091
Formula Weight 203.63
Physical Form Powder/Solid
Chemical Name or Material Pyridoxal Hydrochloride

Recommended Storage : 0°C, desiccate, protect from light.